  • Search Results
  • [Direct involvement of the glucose and Mg(2+)-ATP in the regulation of the GABAA-coupled Cl-,H(CO3(-) -activated Mg(2+)-ATPase of the plasma membrane from rat brain in vitro experiences].
[Direct involvement of the glucose and Mg(2+)-ATP in the regulation of the GABAA-coupled Cl-,H(CO3(-) -activated Mg(2+)-ATPase of the plasma membrane from rat brain in vitro experiences].
Patologicheskaia fiziologiia i eksperimental'naia terapiia (2013-06-29)
S A Menzikov, M N Karpova, M V Kalinina
ABSTRACT
Effect of the glucose and Mg(2+)-ATP on the coupled with CABA(A)-receptor Cl-,HCO3(-)-activated Mg(2+)-ATPase of the plasma membranes from rat brain involving from "basal" Mg(2+)-ATPase which it is activated by Cl-,HCO3(-) ions it was investigated. The glucose (1-10 mM) decreased the "basal" Mg(2+)-ATPase activity on 17% and completely eliminated the enzyme activation by 10 mM Cl(-)+2 mM HCO3(-) ions. The variety effect of the glucose and Mg(2+)-ATP on the activation of the '"basal" Mg(2+)-ATPase by variety concentrations of anions it was established. So it was found that in the presence Mg(2+)-ATP in the incubation medium > 1 mM the enzyme activation by 10 mM Cl- + 2 mM HCO ions not appear. However, in the presence of the glucose (10 mM) the inhibiting effect of the Mg(2+)-ATP on the enzyme is disappears and Cl-,HCO3(-)-ATPase activity is restored. While, in the presence of the high concentrations 40 mM Cl- + 8 mM HCO3(-) in the incubation medium the inhibiting effect of the glucose and Mg(2+)-ATP it was negligible (-20%). It was conclusion about direct involvement of the glucose and Mg(2+)-ATP in the regulation of the coupled with CABA(A)-receptor Cl-,HCO3(-)-ATPase activity of the neuronal membrane under different concentration anions in the incubation medium.
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%