  • Search Results
  • Extensive Ace2 duplication and multiple mutations on Ace1 and Ace2 are related with high level of organophosphates resistance in Aphis gossypii.
Extensive Ace2 duplication and multiple mutations on Ace1 and Ace2 are related with high level of organophosphates resistance in Aphis gossypii.
Environmental toxicology (2012-04-11)
Qingli Shang, Yiou Pan, Kui Fang, Jinghui Xi, Andrew Wong, James Andrew Brennan, Chuanwang Cao
ABSTRACT
Aphis gossypii (Glover) has been found to possess multiple mutations in the acetylcholinesterase (AChE) gene (Ace) that might involve target site insensitivity. In vitro functional expression of AChEs reveals that the resistant Ace1 (Ace1R) and Ace2 (Ace2R) were significantly less inhibited by eserine, omethoate, and malaoxon than the susceptible Ace1 (Ace1S) and Ace2 (Ace2S). Furthermore, in both the mutant and susceptible AChEs, Ace2 was significantly less sensitive to eserine, omethoate, and malaoxon than Ace1. These results suggested that both the mutant Ace1 and Ace2 were responsible for omethoate resistance, while the mutant Ace2 played a major role in insecticide resistance. The DNA copy number and transcription level of Ace2 were 1.52- and 1.88-fold higher in the ORR strain than in the OSS strain. Furthermore, the DNA copy number and transcription level of Ace2 were significantly higher than that of Ace1 in either OSS or ORR strains, demonstrating the involvement of Ace2 gene duplication in resistance. Thus, the authors conclude that omethoate resistance in cotton aphids appears to have evolved through a combination of multiple mutations and extensive Ace2R gene duplication.
Product Description
Supelco
Omethoate, PESTANAL®, analytical standard
Malathion impurity B, European Pharmacopoeia (EP) Reference Standard
Supelco
Malathion, PESTANAL®, analytical standard
Supelco
Malaoxon, PESTANAL®, analytical standard
Supelco
Malathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard
Supelco
Malathion, certified reference material, TraceCERT®
Malathion, European Pharmacopoeia (EP) Reference Standard