  • Search Results
  • Dynamic expression of microRNAs in M2b polarized macrophages associated with systemic lupus erythematosus.
Dynamic expression of microRNAs in M2b polarized macrophages associated with systemic lupus erythematosus.
Gene (2014-07-06)
Pengli Xiao, Chunsheng Dong, Yan Yue, Sidong Xiong
ABSTRACT
Macrophage polarization contributes to the initiation and perpetuation of systemic lupus erythematosus (SLE). Our previous study demonstrated that M2b polarized macrophages induced by activated lymphocyte-derived DNA (ALD-DNA) have a crucial function in the initiation and progress of SLE disease. Accumulated data suggest that microRNAs (miRNAs) serve as critical regulators to control macrophage polarization. To investigate miRNA regulation during macrophage M2b polarization of SLE, miRNA microarrays of murine bone marrow derived macrophages (BMDMs) were performed following stimulation with ALD-DNA for 6 and 36 h. Over 11% of the 1111 analyzed miRNAs appeared differentially expressed during ALD-DNA triggered macrophage M2b polarization. Cluster analysis revealed certain patterns in miRNA expression that are closely linked to ALD-DNA induced macrophage M2b polarization. Analysis of the network structure showed that the predicted functions of the differentially regulated miRNAs at 6h are significantly associated with inflammatory response and disease. Differentially regulated miRNAs identified at 36 h were determined to be significantly related to cell proliferation by biological network analysis. In this study, dynamic miRNA expression patterns and network analysis are described for the first time during ALD-DNA induced macrophage M2b polarization. The data not only provide a better understanding of miRNA-mediated macrophage polarization but also demonstrate the future therapeutic potential of targeting miRNAs in SLE patients.
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from human placenta, Type XIII
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Deoxyribonucleic acid from Micrococcus luteus (lysodeikticus), Type XI, Highly polymerized
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization