  • Search Results
  • N(1)-methylnicotinamide as an endogenous probe for drug interactions by renal cation transporters: studies on the metformin-trimethoprim interaction.
N(1)-methylnicotinamide as an endogenous probe for drug interactions by renal cation transporters: studies on the metformin-trimethoprim interaction.
European journal of clinical pharmacology (2015-01-02)
Fabian Müller, Constanza A Pontones, Bertold Renner, Maren Mieth, Eva Hoier, Daniel Auge, Renke Maas, Oliver Zolk, Martin F Fromm
ABSTRACT
N(1)-methylnicotinamide (NMN) was proposed as an in vivo probe for drug interactions involving renal cation transporters, which, for example, transport the oral antidiabetic drug metformin, based on a study with the inhibitor pyrimethamine. The role of NMN for predicting other interactions with involvement of renal cation transporters (organic cation transporter 2, OCT2; multidrug and toxin extrusion proteins 1 and 2-K, MATE1 and MATE2-K) is unclear. We determined inhibition of metformin or NMN transport by trimethoprim using cell lines expressing OCT2, MATE1, or MATE2-K. Moreover, a randomized, open-label, two-phase crossover study was performed in 12 healthy volunteers. In each phase, 850 mg metformin hydrochloride was administered p.o. in the evening of day 4 and in the morning of day 5. In phase B, 200 mg trimethoprim was administered additionally p.o. twice daily for 5 days. Metformin pharmacokinetics and effects (measured by OGTT) and NMN pharmacokinetics were determined. Trimethoprim inhibited metformin transport with K i values of 27.2, 6.3, and 28.9 μM and NMN transport with IC50 values of 133.9, 29.1, and 0.61 μM for OCT2, MATE1, and MATE2-K, respectively. In the clinical study, trimethoprim increased metformin area under the plasma concentration-time curve (AUC) by 29.5 % and decreased metformin and NMN renal clearances by 26.4 and 19.9 %, respectively (p ≤ 0.01). Moreover, decreases of NMN and metformin renal clearances due to trimethoprim correlated significantly (r S=0.727, p=0.010). These data on the metformin-trimethoprim interaction support the potential utility of N(1)-methylnicotinamide as an endogenous probe for renal drug-drug interactions with involvement of renal cation transporters.
Sigma-Aldrich
β-Nicotinamide mononucleotide, ≥95% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥98% (HPLC)
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Trimethoprim, crystallized, ≥99.0% (HPLC)
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Metformin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Trimethoprim for system suitability, European Pharmacopoeia (EP) Reference Standard