  • Search Results
  • Development of an LC-MS/MS method for high throughput quantification of metformin uptake in transporter inhibition assays.
Development of an LC-MS/MS method for high throughput quantification of metformin uptake in transporter inhibition assays.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-16)
Marianne Vath, Lizbeth Gallagher, Wilson Shou, Harold Weller, Lisa Elkin, Jun Zhang
ABSTRACT
A high throughput LC-MS/MS method for quantification of metformin substrate uptake enables conversion of radiometric transporter inhibition assays for multidrug and toxin extrusion transporters (MATE 1 and 2) and organic cation transporter 2 (OCT2) to a nonradioactive format. Such conversion greatly simplifies assay complexity and reduces assay costs. The development of a quantitative LC-MS/MS method for metformin in support of the high throughput transporter inhibition assays faced specific challenges of achieving both adequate chromatographic retention and rapid analytical turnaround. Here we report a method that circumvents both challenges. The utilization of a porous graphitic carbon column (Hypercarb) ensured adequate retention of highly polar metformin in biological samples. The combined employment of a ballistic gradient on a 3 mm × 30 mm, 5 μm Hypercarb column, and dual staggered chromatography coupled with multiple injection chromatography acquisition, yielded a fast injection-to-injection cycle time of 30s. The method demonstrated good accuracy, precision and excellent robustness for high throughput applications, and has been successfully implemented in the development and validation of the nonradioactive transporter inhibition assays for MATEs and OCT2.
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%