  • Search Results
  • Comprehensive quantitative analysis of Shuang-Huang-Lian oral liquid using UHPLC-Q-TOF-MS and HPLC-ELSD.
Comprehensive quantitative analysis of Shuang-Huang-Lian oral liquid using UHPLC-Q-TOF-MS and HPLC-ELSD.
Journal of pharmaceutical and biomedical analysis (2014-09-16)
Tian-Bo Zhang, Rui-Qi Yue, Jun Xu, Hing-Man Ho, Dik-Lung Ma, Chung-Hang Leung, Siu-Leung Chau, Zhong-Zhen Zhao, Hu-Biao Chen, Quan-Bin Han
ABSTRACT
Shuang-Huang-Lian oral liquid (SHL) is a well-known Chinese patent drug containing three herbal medicines: Radix Scutellariae, Flos Lonicerae Japonicae and Fructus Forsythiae. It is usually used to treat acute upper respiratory tract infection caused by virus or bacteria. Although the licensing of botanical drug Veregen approved by FDA has indicated the importance of quantitative analysis in quality control of herbal medicines, quantitative evaluation of a Chinese patent drug like SHL remains a challenge due to the complex chemical profile. In this study, 15 small molecular components of SHL (four flavonoids, six quinic acid derivatives, three saponins and two phenylethanoid glycosides) were simultaneously determined using ultra-high performance liquid chromatography coupled with quadrupole-time-of-flight mass spectrometry (UHPLC-Q-TOF-MS). The contents of the three major saccharides, namely fructose, glucose and sucrose were quantified using high performance liquid chromatography-evaporative light scattering detector on an amino column (HPLC-ELSD). The macromolecules were quantified by precipitating in 80% ethanol, drying the precipitate, and then weighing. The established methods were validated in terms of linearity, sensitivity, precision, accuracy and stability and then successfully applied to analyze 12 batches of commercial products of SHL produced by four different manufacturers. The results indicated that 57.52-78.11% (w/w) of SHL could be quantitatively determined (non-saccharide small molecules: 1.77-3.75%, monosaccharides: 0.93-20.93%, macromolecules: 2.63-5.76% and sucrose: 49.20-65.94%). This study may provide a useful way to comprehensively evaluate the quality of SHL.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Supelco
Acetonitrile, analytical standard
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sucrose, European Pharmacopoeia (EP) Reference Standard