  • Search Results
  • Prolonged therapy with the soluble guanylyl cyclase activator BAY 60-2770 restores the erectile function in obese mice.
Prolonged therapy with the soluble guanylyl cyclase activator BAY 60-2770 restores the erectile function in obese mice.
The journal of sexual medicine (2014-09-10)
Fábio H Silva, Luiz O Leiria, Eduardo C Alexandre, Ana Paula C Davel, Fabíola Z Mónica, Gilberto De Nucci, Edson Antunes
ABSTRACT
Cardiovascular and endocrine-metabolic diseases associated with increased oxidative stress such as obesity lead to erectile dysfunction (ED). Activators of soluble guanylyl cyclase (sGC) such as BAY 60-2770 reactivate the heme-oxidized sGC in vascular diseases. This study aimed to evaluate the effects of 2-week oral intake with BAY 60-2270 on a murine model of obesity-associated ED. C57BL/6 male mice were fed for 12 weeks with standard chow or high-fat diet. Lean and obese mice were treated with BAY 60-2770 (1 mg/kg/day, 2 weeks). Measurements of intracavernosal pressure (ICP), along with acetylcholine (10(-9) to 10(-5)  M) and electrical field stimulation (EFS; 4-10 Hz)-induced corpus cavernosum relaxations in vitro, were obtained. Levels of cyclic guanosine monophosphate (cGMP), reactive oxygen species (ROS), and sGC protein expressions in cavernosal tissues were measured. Cavernous nerve stimulation caused frequency-dependent ICP increases, which were significantly lower in obese compared with lean mice (P < 0.05). Two-week therapy with BAY 60-2770 fully reversed the decreased ICP in obese group. Acetylcholine-induced cavernosal relaxations were 45% lower (P < 0.001) in obese mice, which were fully restored by BAY 60-2770 treatment. Likewise, the EFS-induced relaxations in obese mice were restored by BAY 60-2770. Basal cGMP content in erectile tissue was 68% lower (P < 0.05) in obese mice, an effect normalized by BAY 60-2770. Levels of ROS were 52% higher (P < 0.05) whereas protein expression of α1 sGC subunit was reduced in cavernosal tissue of obese mice, both of which were normalized by BAY 60-2770. In lean group, BAY 60-2770 did not significantly affect any functional, biochemical, or molecular parameter analyzed. Two-week therapy with BAY 60-2770 restores the erectile function in obese mice that is associated with reduced ROS levels, up-regulation of α1 sGC subunit, and increased cGMP levels in the erectile tissue.
Sigma-Aldrich
3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, AldrichCPR
Sigma-Aldrich
Atropine, ≥99% (TLC), powder
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Phenylephrine, European Pharmacopoeia (EP) Reference Standard