  • Search Results
  • Association between changes in reproductive activity and D-glucose metabolism in the tephritid fruit fly, Bactrocera dorsalis (Hendel).
Association between changes in reproductive activity and D-glucose metabolism in the tephritid fruit fly, Bactrocera dorsalis (Hendel).
Scientific reports (2014-12-17)
Daifeng Cheng, Langjie Chen, Chunyan Yi, Guangwen Liang, Yijuan Xu
ABSTRACT
Reproduction is an important life process in insects; however, few studies have attempted to demonstrate the association between reproductive activity and energy metabolism. To address this problem, we focused on the reproductive changes in Bactrocera dorsalis males. We analyzed B. dorsalis male gene expression profiles during mating (DM), 3 h after mating (A3HM) and 12 h after mating (A12HM). Gene annotation and pathway analyses of differentially expressed genes show that galactose metabolism and the starch and sucrose metabolism pathway activities were significantly higher in A12HM group. Moreover, the maltase D gene was the most strongly up-regulated gene. The D-glucose levels were significantly higher in A12HM group. Maltase D expression level was significantly higher in males reared with sucrose. Body weights of the males reared with D-glucose and sucrose were significantly higher than those of the males reared with yeast extract. We observed more mated males from the groups fed sucrose and D-glucose than from those fed yeast extract. The D-glucose levels in individual males were highest at 18:00 h, when flies exhibit the most active mating behavior. This study shows that the maltase D gene and D-glucose are the critical gene and substrate, respectively, in male B. dorsalis mating process.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material