  • Search Results
  • Immunological shielding by induced recruitment of regulatory T-lymphocytes delays rejection of islets transplanted in muscle.
Immunological shielding by induced recruitment of regulatory T-lymphocytes delays rejection of islets transplanted in muscle.
Cell transplantation (2014-02-01)
Evelina Vågesjö, Gustaf Christoffersson, Tomas B Waldén, Per-Ola Carlsson, Magnus Essand, Olle Korsgren, Mia Phillipson
ABSTRACT
The only clinically available curative treatment of type 1 diabetes mellitus is replacement of the pancreatic islets by allogeneic transplantation, which requires immunosuppressive therapies. Regimens used today are associated with serious adverse effects and impaired islet engraftment and function. The aim of the current study was to induce local immune privilege by accumulating immune-suppressive regulatory T-lymphocytes (Tregs) at the site of intramuscular islet transplantation to reduce the need of immunosuppressive therapy during engraftment. Islets were cotransplanted with a plasmid encoding the chemokine CCL22 into the muscle of MHC-mismatched mice, after which pCCL22 expression and leukocyte recruitment were studied in parallel with graft functionality. Myocyte pCCL22 expression and secretion resulted in local accumulation of Tregs. When islets were cotransplanted with pCCL22, significantly fewer effector T-lymphocytes were observed in close proximity to the islets, leading to delayed graft rejection. As a result, diabetic recipients cotransplanted with islets and pCCL22 intramuscularly became normoglycemic for 10 consecutive days, while grafts cotransplanted with control plasmid were rejected immediately, leaving recipients severely hyperglycemic. Here we propose a simple method to initially shield MHC-mismatched islets by the recruitment of endogenous Tregs during engraftment in order to improve early islet survival. Using this approach, the very high doses of systemic immunosuppression used initially following transplantation can thereby be avoided.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Methyl salicylate, analytical standard
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
Methyl salicylate, BioXtra
Supelco
Methyl salicylate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)