  • Search Results
  • Saikosaponin-d affects the differentiation, maturation and function of monocyte-derived dendritic cells.
Saikosaponin-d affects the differentiation, maturation and function of monocyte-derived dendritic cells.
Experimental and therapeutic medicine (2014-06-19)
Zuo-Lin Ying, Xiao-Jie Li, Hong Dang, Feng Wang, Xiao-Yan Xu
ABSTRACT
Saikosaponin-d (Ssd) is a triterpenoid saponin derived from
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%