  • Search Results
  • Ultrathin Nanotube/Nanowire Electrodes by Spin-Spray Layer-by-Layer Assembly: A Concept for Transparent Energy Storage.
Ultrathin Nanotube/Nanowire Electrodes by Spin-Spray Layer-by-Layer Assembly: A Concept for Transparent Energy Storage.
ACS nano (2015-09-08)
Forrest S Gittleson, Daniel Hwang, Won-Hee Ryu, Sara M Hashmi, Jonathan Hwang, Tenghooi Goh, André D Taylor
ABSTRACT
Fully integrated transparent devices require versatile architectures for energy storage, yet typical battery electrodes are thick (20-100 μm) and composed of optically absorbent materials. Reducing the length scale of active materials, assembling them with a controllable method and minimizing electrode thickness should bring transparent batteries closer to reality. In this work, the rapid and controllable spin-spray layer-by-layer (SSLbL) method is used to generate high quality networks of 1D nanomaterials: single-walled carbon nanotubes (SWNT) and vanadium pentoxide (V2O5) nanowires for anode and cathode electrodes, respectively. These ultrathin films, deposited with ∼2 nm/bilayer precision are transparent when deposited on a transparent substrate (>87% transmittance) and electrochemically active in Li-ion cells. SSLbL-assembled ultrathin SWNT anodes and V2O5 cathodes exhibit reversible lithiation capacities of 23 and 7 μAh/cm(2), respectively at a current density of 5 μA/cm(2). When these electrodes are combined in a full cell, they retain ∼5 μAh/cm(2) capacity over 100 cycles, equivalent to the prelithiation capacity of the limiting V2O5 cathode. The SSLbL technique employed here to generate functional thin films is uniquely suited to the generation of transparent electrodes and offers a compelling path to realize the potential of fully integrated transparent devices.
Product Description
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Sigma-Aldrich
Ethylene oxide solution, 2.5-3.3 M in THF
Supelco
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
Sigma-Aldrich
Vanadium, foil, thickness 0.5 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 1.0 mm, 99.7%
Sigma-Aldrich
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis