  • Search Results
  • Metastasis of breast cancer cells to the bone, lung, and lymph nodes promotes resistance to ionizing radiation.
Metastasis of breast cancer cells to the bone, lung, and lymph nodes promotes resistance to ionizing radiation.
Strahlentherapie und Onkologie : Organ der Deutschen Rontgengesellschaft ... [et al] (2017-06-24)
Takamitsu Hara, Manabu Iwadate, Kazunoshin Tachibana, Satoshi Waguri, Seiichi Takenoshita, Nobuyuki Hamada
ABSTRACT
Metastasis represents the leading cause of breast cancer deaths, necessitating strategies for its treatment. Although radiotherapy is employed for both primary and metastatic breast cancers, the difference in their ionizing radiation response remains incompletely understood. This study is the first to compare the radioresponse of a breast cancer cell line with its metastatic variants and report that such metastatic variants are more radioresistant. A luciferase expressing cell line was established from human basal-like breast adenocarcinoma MDA-MB-231 and underwent in vivo selections, whereby a cycle of inoculations into the left cardiac ventricle or the mammary fat pad of athymic nude mice, isolation of metastases to the bone, lung and lymph nodes visualized with bioluminescence imaging, and expansion of obtained cells was repeated twice or three times. The established metastatic cell lines were assessed for cell proliferation, wound healing, invasion, clonogenic survival, and apoptosis. The established metastatic cell lines possessed an increased proliferative potential in vivo and were more chemotactic, invasive, and resistant to X‑ray-induced clonogenic inactivation and apoptosis in vitro. Breast cancer metastasis to the bone, lung, and lymph nodes promotes radioresistance.
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture