Merck
All Photos(1)

775002

Deuterium oxide

99.96 atom % D, potassium phosphate buffer 80 mM, contains 2 mM DSS-d6 (98 atom % D), pH 7

Synonym(s):
Heavy water, Water-d2
Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
Número MDL:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

pureza isotópica

99.96 atom % D

contém

2 mM DSS-d6 (98 atom % D)

composição

potassium phosphate buffer, 80 mM

technique(s)

NMR: suitable

pH

7

pb

101.4 °C (lit.)

pf

3.8 °C (lit.)

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Embalagem

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service