Merck
All Photos(2)
802662
Sigma-Aldrich
Aquivion® P98-SO2F
cylinder (pellet), size 2 mm, PFSF eq. wt. (980 g/mole SO2F)
Synonym(s):
Aquivion® SO2F-form, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid
Linear Formula:
[CF2CF(OCF2CF2SO2F)]m[CF2CF2]n
CAS Number:
NACRES:
NA.23
forma
cylinder (pellet)
pellets
Peso eq.
980±40g/mol (SO2F)
tamanho
2 mm
Looking for similar products? Visit Product Comparison Guide
Descrição geral
Aquivion® P98-SO2F are pellets based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) CF2=CF2-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. The resulting perfluoropolymer′s functional groups are in their sulfonyl fluoride form, SO2F.
Aplicação
Aquivion® P98 pellets can be melt-pressed or extruded into plates, sheets or thin films down to about 50μ. The extruder shall be equipped with a standard three-zone metering screw devoid of barrier or mixing elements. Optimum temperature settings will depend on the configuration of the equipment but generally cover 100 to 300°C range. Optionally functional groups in the molded part are then hydrolyzed in baths or wet benches with a heated base (NaOH, KOH etc), reaction which may take minutes or hours, depending on thickness and conditions.
Aquivion® P98 pellets can be directly hydrolyzed as described - either completely or only on their outer surface. This will enable functional activity of the sulfonic acid /sulfonyl salt groups for ion-exchange operations or as acid catalyst.
Informações legais
Aquivion is a registered trademark of Solvay Group
Código de classe de armazenamento
11 - Combustible Solids
WGK
WGK 3
Ponto de fulgor (ºF)
Not applicable
Ponto de fulgor (ºC)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service