Pular para o conteúdo
Merck

941174

NanoFabTx photoactivatable lipid mix

AzoPC, for small molecule delivery

Faça login para ver os preços organizacionais e de contrato.

Selecione um tamanho

Preço e disponibilidade não estão disponíveis no momento.

Pular para

Assistência Técnica
Precisa de ajuda? Nossa equipe de cientistas experientes está à sua disposição.
Estamos aqui para ajudar

description

Technique(s): microfluidics, extrusion, flash nanoprecipitation

form

(powder, chunks, sold)

composition

AzoPC , Contains: DSPC, Cholesterol, AzoPC

color

white to off-white

suitability

suitable for ( small molecule delivery)

functional group

(Photoswitchable AzoPC)

storage temp.

−20°C

Quality Level

General description

About the material:

NanoFabTx AzoPC Lipid Mix is a curated lipid mixture designed for the synthesis of photoactivatable liposomes. This ready-to-use lipid mix contains carefully selected lipids and comes with detailed step-by-step instructions for the synthesis process.

The photoactivatable lipid AzoPC exhibits cytotoxicity upon exposure to UV-A light in human cancer cell lines, leading to the release of its payload upon photoactivation. Notably, the photoactivatable liposomes themselves are non-cytotoxic in the absence of light.

For further details and a comprehensive step-by-step protocol, please refer to the "More Documents" section on this page.

Application

The NanoFabTx photoactivatable AzoPC lipid mix can be used to:

  • Enable the synthesis of photoactivatable liposomes for targeted drug delivery
  • Allow controlled release of payload upon photoactivation
  • Research and development in the field of targeted cancer therapy and drug delivery technologies

Features and Benefits

  • A ready-to-use lipid blend for the synthesis of photoactivatable liposomes.
  • Step-by-step protocols (microfluidics) developed and tested by our formulation scientists
  • Flexible synthesis tool to create uniform and reproducible liposomes
Comprehensive protocols for liposome synthesis are included:
  • A microfluidics protocol using commercial platforms or syringe pumps
The microfluidics protocol included with this product uses the NanoFabTx device kit (911593). These kits come with the microfluidics chips, fittings, and tubing required to get started with microfluidics-based synthesis (compatible microfluidics system or syringe pump required).

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Escolha uma das versões mais recentes:

Certificados de análise (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documentos section.

Se precisar de ajuda, entre em contato Atendimento ao cliente

Já possui este produto?

Encontre a documentação dos produtos que você adquiriu recentemente na biblioteca de documentos.

Visite a Biblioteca de Documentos

Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.

Entre em contato com a assistência técnica