Merck
  • GF46978385
All Photos(2)

GF46978385

Iron

foil, not light tested, 100x100mm, thickness 0.0125mm, 99.85%

Synonym(s):
Iron, FE000160
Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
Número MDL:
ID de substância PubChem:
NACRES:
NA.23

Ensaio

99.85%

forma

foil

manufacturer/tradename

Goodfellow 469-783-85

resistividade

9.71 μΩ-cm

tamanho × espessura

100 x 100 mm × 0.0125 mm

pb

2750 °C (lit.)

pf

1535 °C (lit.)

densidade

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

1 of 4

This Item
GF52626607GF78366156GF81988240
Iron foil, not light tested, 100x100mm, thickness 0.0125mm, 99.85%

GF46978385

Iron

Iron foil, not light tested, 100x100mm, thickness 0.0025mm, 99.85%

GF52626607

Iron

Iron foil, not light tested, 100x100mm, thickness 0.015mm, 99.85%

GF78366156

Iron

Iron foil, not light tested, 100x100mm, thickness 0.003mm, 99.85%

GF81988240

Iron

assay

99.85%

assay

99.85%

assay

99.85%

assay

99.85%

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 469-783-85

manufacturer/tradename

Goodfellow 526-266-07

manufacturer/tradename

Goodfellow 783-661-56

manufacturer/tradename

Goodfellow 819-882-40

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

Descrição geral

For updated SDS information please visit www.goodfellow.com.

Informações legais

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service