GF47677012

Palladium

foil, 25mm disks, thickness 0.008mm, 99.95%

Sinônimo(s):
PD000240, Palladium
Fórmula linear:
Pd
Número CAS:
Peso molecular:
106.42
Número MDL:
ID de substância PubChem:
Preço e disponibilidade não estão disponíveis no momento.

teor

99.95%

forma

foil

Nº de fabricação

Goodfellow, 476-770-12

resistividade

9.96 μΩ-cm, 20°C

diâmetro × espessura

25 mm × 0.008 mm

pb

2970 °C (lit.)

pf

1554 °C (lit.)

densidade

12.02 g/cm3 (lit.)

SMILES string

Pd

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Procurando produtos similares? Visit Guia de comparação de produtos

Descrição geral

For updated SDS information please visit www.goodfellow.com.

Informações legais

Product of Goodfellow

WGK Alemanha

nwg

Ponto de fulgor (ºF)

Not applicable

Ponto de fulgor (ºC)

Not applicable

Kiyofumi Inamoto
Chemical & pharmaceutical bulletin, 61(10), 987-996 (2013-10-04)
Herein, we describe our development of synthetic methods for heterocyclic compounds based on the palladium-catalyzed carbon-hydrogen bond (C-H) functionalization/intramolecular carbon-heteroatom (nitrogen or sulfur) bond formation process. By this C-H cyclization method, we efficiently prepared various N-heterocycles, including indazoles, indoles, and...
Qing-An Chen et al.
Chemical Society reviews, 42(2), 497-511 (2012-11-10)
The transition metal catalyzed asymmetric hydrogenation of unsaturated compounds arguably presents one of the most attractive methods for the synthesis of chiral compounds. Over the last few decades, Pd has gradually grown up as a new and popular metal catalyst...
Xiao-Feng Wu et al.
ChemSusChem, 6(2), 229-241 (2013-01-12)
Palladium-catalyzed coupling reactions have become a powerful tool for advanced organic synthesis. This type of reaction is of significant value for the preparation of pharmaceuticals, agrochemicals, as well as advanced materials. Both, academic as well as industrial laboratories continuously investigate...
Stephan Enthaler et al.
Chemical Society reviews, 40(10), 4912-4924 (2011-06-07)
The formation of oxygen-carbon bonds is one of the fundamental transformations in organic synthesis. In this regard the application of palladium-based catalysts has been extensively studied during recent years. Nowadays it is an established methodology and the success has been...
Abdellatif Essoumhi et al.
Journal of nanoscience and nanotechnology, 14(2), 2012-2023 (2014-04-23)
This review reports on the recent advances in the synthesis and physico-chemical properties of palladium-containing perovskites. Initially, the perovskite structure is briefly reviewed, then palladium-containing perovskites synthesis and physico-chemical properties are detailed. The applications of palladium-containing perovskites in catalysis; namely...

Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.

Entre em contato com a assistência técnica

Redes sociais

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Pesquisa. Desenvolvimento. Produção.

Somos um fornecedor global líder para o setor de ciências biológicas, com soluções e serviços para pesquisa, desenvolvimento e produção de biotecnologia, além de produção e desenvolvimento de terapias farmacológicas com medicamentos.

© 2021 Merck KGaA, Darmstadt, Alemanha e/ou suas filiais. Todos os direitos reservados.

A reprodução não autorizada de quaisquer materiais deste site é estritamente proibida.