Todas as fotos(2)

GF80082479

Gold

sphere, 200pcs, diameter 0.5mm, 99.99%

Sinônimo(s):
Gold, AU006840
Empirical Formula (Hill Notation):
Au
Número CAS:
Peso molecular:
196.97
Número MDL:
ID de substância PubChem:

teor

99.99%

forma

spheres

manufacturer/tradename

Goodfellow 800-824-79

resistividade

2.05 μΩ-cm, 0°C

tamanho de partícula

0.5 mm

pb

2808 °C (lit.)

pf

1063 °C (lit.)

densidade

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Categorias relacionadas

Descrição geral

For updated SDS information please visit www.goodfellow.com.

Informações legais

Product of Goodfellow

Código de classe de armazenamento

11 - Combustible Solids

WGK

nwg

Ponto de fulgor (ºF)

Not applicable

Ponto de fulgor (ºC)

Not applicable

Certificado de análise

Certificado de origem

Lin Luo et al.
Journal of nanoscience and nanotechnology, 14(6), 4036-4042 (2014-04-18)
A novel immunosensor of human immune globulin (IgG) was fabricated based on the fluorescence transfer between luminescent semiconductor quantum dots (QDs) and gold nanoparticles (AuNPs). AuNPs and CdSe/ZnS QDs were respectively labeled with immune reaction pair:IgG and goat anti-human immunoglobulin
Jiajing Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4124-4138 (2014-04-18)
Gold nanostructures with promising applications in biomedical field have attracted great attention. However, some fundamental questions other than the development of novel applications should be elucidated before they can actually serve as biomedicines in the clinic. Bio-safety is one of
Maria Jose Santos-Martinez et al.
Journal of biomedical nanotechnology, 10(6), 1004-1015 (2014-04-23)
The increasing use of gold nanoparticles in medical diagnosis and treatment has raised the concern over their blood compatibility. The interactions of nanoparticles with blood components may lead to platelet aggregation and endothelial dysfunction. Therefore, medical applications of gold nanoparticles
Cathleen M Crudden et al.
Nature chemistry, 6(5), 409-414 (2014-04-24)
Since the first report of thiol-based self-assembled monolayers (SAMs) 30 years ago, these structures have been examined in a huge variety of applications. The oxidative and thermal instabilities of these systems are widely known, however, and are an impediment to
Guojun Weng et al.
Journal of nanoscience and nanotechnology, 14(6), 4072-4077 (2014-04-18)
Gold nanorods of different aspect ratios had been synthesized using seed mediated growth method. The formed gold nanorods had been characterized by the absorption and transmission electron microscopy (TEM) measurements. The obtained gold nanorods were used to study the quenched

Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.

Entre em contato com a assistência técnica