Selecione um tamanho
R$ 2.438,00
R$ 2.438,00
Sobre este item
Pular para
material
borosilicate glass , round bottom flask, three-neck (Vertical)
joint
Joints: ST/NS24/40 (3)
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Ace Glass 6944-60
capacity
5,000 mL
Categorias relacionadas
1 of 4
Este Item | Z125784 | Z125652 | Z125687 |
|---|---|---|---|
| manufacturer/tradename Ace Glass 6944-60 | manufacturer/tradename Ace Glass 6948-58 | manufacturer/tradename Ace Glass 6944-18 | manufacturer/tradename Ace Glass 6944-48 |
| material borosilicate glass , three-neck (Vertical), round bottom flask | material borosilicate glass , round bottom flask, three angled neck | material borosilicate glass flask, round bottom flask, three-neck (Vertical) | material borosilicate glass , round bottom flask, three-neck (Vertical) |
| capacity 5,000 mL | capacity 5,000 mL | capacity 500 mL | capacity 3,000 mL |
| joint Joints: ST/NS24/40 (3) | joint Joints: ST/NS24/40 (3) | joint center joint: ST/NS29/42, side Joints: ST/NS24/40 (2) | joint Joints: ST/NS24/40 (3) |
| packaging pack of 1 | packaging pack of 1 | packaging pack of 1 | packaging pack of 1 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
Escolha uma das versões mais recentes:
Já possui este produto?