Merck
All Photos(1)

37744

Supelco

AQUANAL-professional Phosphate total tube test

Número EC:

measuring range

5.0-60 mg/L

Nível de qualidade

100

application(s)

environmental
general analytical

Descrição geral

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL-professional tube tests were developed to provide a fast safe and clean method in order to perform water analysis in sewage treatment plants. The sample is pipetted into a tube that contains the test reagent. For some tests, an additional reagent is required that has to be added manually.

Embalagem

24 reaction tubes, 1 blank tube

Informações legais

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Palavra indicadora

Danger

Classificações de perigo

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Órgãos-alvo

Respiratory system

Código de classe de armazenamento

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Ponto de fulgor (ºF)

Not applicable

Ponto de fulgor (ºC)

Not applicable

Equipamento de proteção individual

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P1 (EN143) respirator filter, type P3 (EN 143) respirator cartridges

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service