Selecione um tamanho
Sobre este item
InChI key
HVYWMOMLDIMFJA-DPAQBDIFSA-N
InChI
1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1
SMILES string
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
biological source
synthetic
assay
≥98% (GC)
form
powder
packaging
pkg of 1 × 1 g IA00001-1G
color
white to off-white
bp
360 °C (lit.)
mp
147-149 °C (lit.), 147-150
solubility
ethanol: soluble, soluble
density
1.067 g/mL at 25 °C (lit.)
shipped in
dry ice
storage temp.
−20°C
Quality Level
Procurando produtos similares? Visita Guia de comparação de produtos
General description
1.Drug Development: Cholesterol analogs are used in the development of drugs targeting cholesterol metabolism and related diseases such as atherosclerosis.
2.Biological Studies: Synthetic cholesterol is utilized to study the effects of cholesterol on cells, cellular processes, and as a standard in biological experiments.
3.Vaccine Development: It is used in the formulation of lipid-based adjuvants for vaccine development due to its ability to enhance immune responses.
4.Drug Delivery System Research: Cholesterol derivatives are incorporated into drug delivery systems to enhance the stability and bioavailability of pharmaceutical compounds.
5.Membrane Studies: Synthetic cholesterol is used to mimic the properties of cell membranes, aiding in the study of drug interactions and membrane permeability.
6.Disease Modeling: It is employed in disease modeling and in vitro studies to understand the role of cholesterol in various diseases.
Application
Biochem/physiol Actions
Features and Benefits
Legal Information
Classe de armazenamento
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Escolha uma das versões mais recentes:
Já possui este produto?
Encontre a documentação dos produtos que você adquiriu recentemente na biblioteca de documentos.
Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.
Entre em contato com a assistência técnica