Merck
All Photos(1)

PE1400

Supelco

Organonitrogen Pesticides - WS

Proficiency Testing Material

Synonym(s):
SDWA Organonitrogen Pesticides
NACRES:
NA.24

Nível de qualidade

embalagem

ampule of 2 × 2 mL

concentração

in acetone

application(s)

environmental

formato

matrix material

temperatura de armazenamento

2-8°C

Descrição geral

Organonitrogen Pesticides - WS proficiency testing material is produced in accordance with ISO/IEC 17043:2010. Also referred to as Potable Water or Water Supply sample, this sample is manufactured according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The lower criteria from the TNI tables mimic the levels common to drinking water samples.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WS program is designed to meet the quality assurance requirements in compliance with the Safe Drinking Water Act (SDWA), thereby ensuring the analytical competence of your laboratory or drinking water facility.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Water Analysis - Water Supply (WS)

Matrix: Water Supply/Drinking Water

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Organonitrogen Pesticides - WS proficiency testing standard can be found in the reporting packet which will be available to download at PT Portal.

Aplicação

Organonitrogen Pesticides - WS as a proficiency testing material finds applications in the determination of organonitrogen pesticides in drinking water samples.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Embalagem

2 mL

Características e benefícios

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Nota de preparo

Description

  • This PT sample is packaged as a 1.5 mL concentrate.
  • The solvent for this sample is acetone.
  • The PT sample is not preserved.
  • The sample must be stored at 4 °C.

Sample Preparation

  • Fill a 1000 mL Class A Volumetric Flask with 900 mL of organic-free reagent water.
  • Allow the ampule concentrate to equilibrate to room temperature.
  • Open the ampule and pipet exactly 1.0 mL of the concentrate into the flask.
  • Fill to volume and mix well.
  • This represents the sample for analysis.
  • Report results in μg/L.

Armazenamento e estabilidade

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Outras notas

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2022-2023)

Drinking Water
WS22-2 March 23 - May 6, 2022

WS23-2 March 22 – May 5, 2023


Quick-Turn Study (on demand proficiency test that is offereddepending on lot availability)

Informações legais

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetochlor 1-50 μg/L
    Alachlor 2-20 μg/L
    Atrazine 2-20 μg/L
    Bromacil 2-20 μg/L
    Butachlor 2-20 μg/L
    Metolachlor 2-20 μg/L
    Metribuzin 2-20 μg/L
    Molinate 5-50 μg/L
    Siamzine 2-20 μg/L
    See All (9)

Pictogramas

FlameExclamation mark

Palavra indicadora

Danger

Frases de perigo

Classificações de perigo

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Órgãos-alvo

Respiratory system

Perigos de suplementos

Código de classe de armazenamento

3 - Flammable liquids

WGK

WGK 1

Ponto de fulgor (ºF)

1.4 °F - closed cup - Solvent

Ponto de fulgor (ºC)

-17 °C - closed cup - Solvent

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service