Merck
All Photos(1)

55120C

SAFC

Phosphate buffered saline

sterile; 0.1 μm filtered, ready-to-use-bag, liquid

Synonym(s):
PBS
Número MDL:

Nível de qualidade

500

esterilidade

sterile; 0.1 μm filtered

forma

liquid

Impurezas

≤1 EU/mL Endotoxin

pH

7.1-7.5 (25 °C±0.5)

osmolalidade

290-330 mOsm/kg H2O

solubilidade

saline: soluble, colorless (Phosphate Buffered Saline (PBS))

temperatura de armazenamento

15-25°C

Looking for similar products? Visit Product Comparison Guide

Descrição geral

  • Manufactured in an ISO 9001:2015 certified facility
  • A 12-month shelf life from the date of production (with a target of 24 months)
  • Manufactured using Emprove® raw materials (complying with Ph. Eur. and most with USP)
  • Ready-to-use bags with standard MPC/MPX connectors
  • Liquid Solutions MyWay products are intended for bioprocessing and further manufacturing uses

Informações legais

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Código de classe de armazenamento

12 - Non Combustible Liquids

WGK

WGK 1

Ponto de fulgor (ºF)

Not applicable

Ponto de fulgor (ºC)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service