Merck
All Photos(1)
55220C
SAFC
Sodium chloride solution
1 M
Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
Número MDL:
esterilidade
sterile; 0.1 μm filtered
Nível de qualidade
200
concentração
1 M
Impurezas
≤1 EU/mL Endotoxin
condutividade
80.6-98.5 mS/cm at 25 °C±0.5
temperatura de armazenamento
15-25°C
SMILES string
[Na+].[Cl-]
InChI
1S/ClH.Na/h1H;/q;+1/p-1
InChI key
FAPWRFPIFSIZLT-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Descrição geral
  • Manufactured in an ISO 9001:2015 certified facility
  • A 12-month shelf life from the date of production (with a target of 24 months)
  • Manufactured using Emprove® raw materials (complying with Ph. Eur. and most with USP)
  • Ready-to-use bags with standard MPC/MPX connectors
  • Liquid Solutions MyWay products are intended for bioprocessing and further manufacturing uses
Embalagem
  • 55220C-1BNC: 10 x 1L Bag
  • 55220C-200BNC: 200L Bag
  • 55220C-20BNC: 20L Bag
Informações legais
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
Código de classe de armazenamento
12 - Non Combustible Liquids
WGK
WGK 1
Ponto de fulgor (ºF)
Not applicable
Ponto de fulgor (ºC)
Not applicable
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
More Documents
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service