sterile; 0.1 μm filtered
200
1 M
≤1 EU/mL Endotoxin
80.6-98.5 mS/cm at 25 °C±0.5
15-25°C
[Na+].[Cl-]
1S/ClH.Na/h1H;/q;+1/p-1
FAPWRFPIFSIZLT-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
12 - Non Combustible Liquids
WGK 1
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service