GE28-9645-14
Sigma-Aldrich
HiScale 26/20
NACRES:
SB.52
aplicação(ões)
LPLC: suitable
Descrição geral
The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments as well as disassembly and cleaning. The measurement scale on the column makes it easier to predict and reach a predefined packing height, thereby increasing process control as well as reproducibility. Although optimized for high-pressure media, HiScale columns are fully compatible with all applications (and packing methods) currently applicable to XK columns.
Características e benefícios
Based on the well-established line of XK columns and compatible with all methods currently applicable to XK columns.
Adapter QuickLock mechanism, column measurement scale, and ergonomic design, facilitate column handling and increase process control.
Dual adapters, offering high flexibility and dynamic packing heights.
Empty columns with high pressure stability for packing with modern BioProcess media for process development and preparative chromatography.
Provides axial compression of the gel bed and a nonrotating plunger mechanism that expands packing capabilities.
Capability to rinse entire adapter shaft after packing, thereby minimizing risk of bacterial growth after prolonged use.
Certificado de análise
Certificado de origem
Artigos
This page describes efficient column packing and preparation for affinity chromatography of antibodies.
Leia mais
This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.
Leia mais
Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.
Leia mais
Protocolos
This page shows how to separate IgG antibodies by affinity chromatography using Protein G Sepharose 4 Fast Flow from Cytiva.
Leia mais
This page shows how to separate IgG antibodies by affinity chromatography using Protein G Sepharose 4 Fast Flow from Cytiva.
Leia mais
This page describes efficient column packing and preparation for affinity chromatography of GST-tagged proteins using Cytiva products.
Leia mais
This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using Ni Sepharose Excel from Cytiva.
Leia mais
Redes sociais
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Pesquisa. Desenvolvimento. Produção.
Somos um fornecedor global líder para o setor de ciências biológicas, com soluções e serviços para pesquisa, desenvolvimento e produção de biotecnologia, além de produção e desenvolvimento de terapias farmacológicas com medicamentos.
© 2021 Merck KGaA, Darmstadt, Alemanha e/ou suas filiais. Todos os direitos reservados.
A reprodução não autorizada de quaisquer materiais deste site é estritamente proibida.