L5149
Sigma-Aldrich
Labels, phosphorescent
4 × 6 inch sheets
embalagem
pkg of 10 sheets
manufacturer/tradename
Diversified Biotech RAD-10
C × L
10.1 cm × 15.25 cm
Categorias relacionadas
Descrição geral
Label sheets for autoradiograms. Identifying marks are written on the label with an ordinary black pen, then affixed to a membrane or gel (the label is self-adhesive). A clear negative image of the markings is transferred to the autoradiogram, regardless of the length of the exposure.
Embalagem
Package contains ten 4 × 6 inch sheets, which may be cut to size.
Certificado de análise
Certificado de origem