SAB1406601

Sigma-Aldrich

Anti-XRCC1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sinônimo(s):
RCC
NACRES:
NA.41

Nível de qualidade

100

fonte biológica

mouse

forma do anticorpo

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

forma

buffered aqueous solution

peso molecular

antigen ~69.5 kDa

species reactivity

human

aplicação(ões)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable
western blot: 1 μg/mL

conjugado

unconjugated

nº de adesão NCBI

nº de adesão UniProt

enviado em

dry ice

temperatura de armazenamento

−20°C

Gene Information

human ... XRCC1(7515)

Descrição geral

The protein encoded by this gene is involved in the efficient repair of DNA single-strand breaks formed by exposure to ionizing radiation and alkylating agents. This protein interacts with DNA ligase III, polymerase beta and poly (ADP-ribose) polymerase to participate in the base excision repair pathway. It may play a role in DNA processing during meiogenesis and recombination in germ cells. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. (provided by RefSeq)

Imunogênio

XRCC1 (AAH23593.1, 1 a.a. ~ 633 a.a) full-length human protein.

Sequence
MPEIRLRHVVSCSSQDSTHCAENLLKADTYRKWRAAKAGEKTISVVLQLEKEEQIHSVDIGNDGSAFVEVLVGSSAGGAGEQDYEVLLVTSSFMSPSESRSGSNPNRVRMFGPDKLVRAAAEKRWDRVKIVCSQPYSKDSPFGLSFVRFHSPPDKDEAEAPSQKVTVTKLGQFRVKEEDESANSLRPGALFFSRINKTSPVTASDPAGPSYAAATLQASSAASSASPVSRAIGSTSKPQESPKGKRKLDLNQEEKKTPSKPPAQLSPSVPKRPKLPAPTRTPATAPVPARAQGAVTGKPRGEGTEPRRPRAGPEELGKILQGVVVVLSGFQNPFRSELRDKALELGAKYRPDWTRDSTHLICAFANTPKYSQVLGLGGRIVRKEWVLDCHRMRRRLPSQRYLMAGPGSSSEEDEASHSGGSGDEAPKLPQKQPQTKTKPTQAAGPSSPQKPPTPEETKAASPVLQEDIDIEGVQSEGQDNGAEDSGDTEDELRRVAEQKEHRLPPGQEENGEDPYAGSTDENTDSEEHQEPPDLPVPELPDFFQGKHFFLYGEFPGDERRKLIRYVTAFNGELEDYMSDRVQFVITAQEWDPSFEEALMDNPSLAFVRPRWIYSCNEKQKLLPHQLYGVVPQA

forma física

Solution in phosphate buffered saline, pH 7.4

storage_class_code

13 - Non Combustible Solids

WGK Alemanha

WGK 1

Ponto de fulgor (ºF)

Not applicable

Ponto de fulgor (ºC)

Not applicable

Certificado de análise

Certificado de origem

Bárbara Alcaraz Silva et al.
The Journal of biological chemistry, 289(33), 22771-22784 (2014-07-02)
Chromosome ends contain nucleoprotein structures known as telomeres. Damage to chromosome ends during interphase elicits a DNA damage response (DDR) resulting in cell cycle arrest. However, little is known regarding the signaling from damaged chromosome ends (designated here as "TIPs")...

Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.

Entre em contato com a assistência técnica