SAB3500294

Sigma-Aldrich

Anti-MRE11 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sinônimo(s):
Anti-Meiotic recombination 11 homolog, Anti-MRE11A, Anti-MRE11B, Anti-ATLD, Anti-HNGS
Número MDL:
NACRES:
NA.41

fonte biológica

rabbit

Nível de qualidade

100

forma do anticorpo

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

forma

buffered aqueous solution

peso molecular

predicted mol wt 78 kDa

species reactivity

mouse, human, rat

aplicação(ões)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

conjugado

unconjugated

nº de adesão NCBI

nº de adesão UniProt

enviado em

dry ice

temperatura de armazenamento

−20°C

Gene Information

human ... MRE11A(4361)

Imunogênio

MRE11 antibody was raised against a 14 amino acid peptide from near the amino terminus human MRE11.

Características e benefícios

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Ligação

The action of this antibody can be blocked using blocking peptide SBP3500294.

forma física

Supplied in PBS with 0.02% sodium azide.

storage_class_code

12 - Non Combustible Liquids

WGK Alemanha

WGK 2

Ponto de fulgor (ºF)

Not applicable

Ponto de fulgor (ºC)

Not applicable

Certificado de análise

Certificado de origem

Rebecca M Martin et al.
Oncotarget, 5(4), 993-1003 (2014-03-15)
Predictive assays are needed to help optimise treatment in muscle-invasive bladder cancer, where patients can be treated by either cystectomy or radical radiotherapy. Our finding that low tumour MRE11 expression is predictive of poor response to radiotherapy but not cystectomy...
Bárbara Alcaraz Silva et al.
The Journal of biological chemistry, 289(33), 22771-22784 (2014-07-02)
Chromosome ends contain nucleoprotein structures known as telomeres. Damage to chromosome ends during interphase elicits a DNA damage response (DDR) resulting in cell cycle arrest. However, little is known regarding the signaling from damaged chromosome ends (designated here as "TIPs")...

Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.

Entre em contato com a assistência técnica