WH0080142M1

Sigma-Aldrich

Monoclonal Anti-PTGES2 antibody produced in mouse

clone 5B7, purified immunoglobulin, buffered aqueous solution

Sinônimo(s):
Anti-GBF1, Anti-C9orf15, Anti-prostaglandin E synthase 2, Anti-FLJ14038, Anti-PGES2, Anti-MGC11289
Número MDL:

Nível de qualidade

100

fonte biológica

mouse

forma do anticorpo

purified immunoglobulin

antibody product type

primary antibodies

clone

5B7, monoclonal

forma

buffered aqueous solution

species reactivity

human

aplicação(ões)

indirect ELISA: suitable
western blot: 1-5 μg/mL

Isotipo

IgG1κ

conjugado

unconjugated

GenBank® accession no.

nº de adesão UniProt

enviado em

dry ice

temperatura de armazenamento

−20°C

Gene Information

human ... PTGES2(80142)

Descrição geral

The protein encoded by this gene is a membrane-associated prostaglandin E synthase, which catalyzes the conversion of prostaglandin H2 to prostaglandin E2. This protein also has been shown to activate the transcription regulated by a gamma-interferon-activated transcription element (GATE). Multiple transcript variants have been found for this gene. (provided by RefSeq)

Imunogênio

PTGES2 (NP_079348, 270 a.a. ~ 377 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
FGAVEGAVAKYMGAAAMYLISKRLKSRHRLQDNVREDLYEAADKWVAAVGKDRPFMGGQKPNLADLAVYGVLRVMEGLDAFDDLMQHTHIQPWYLRVERAITEASPAH

Ações bioquímicas/fisiológicas

Prostaglandin E synthase 2 (PTGES2) is involved in the formation of prostaglandin E2 from prostaglandin H2.

forma física

Solution in phosphate buffered saline, pH 7.4

Informações legais

GenBank is a registered trademark of United States Department of Health and Human Services

Equipamento de proteção individual

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

RIDADR

NONH for all modes of transport

WGK Alemanha

nwg

Ponto de fulgor (ºF)

Not applicable

Ponto de fulgor (ºC)

Not applicable

Certificado de análise
Certificado de origem
Taro Yamada et al.
Biochemistry, 46(28), 8414-8424 (2007-06-26)
Prostaglandin E2 synthase (PGES) catalyzes the isomerization of PGH2 to PGE2. PGES type 2 (mPGES-2) is a membrane-associated enzyme, whose N-terminal section is apparently inserted into the lipid bilayer. Both intact and N-terminal truncated enzymes have been isolated and have...
Inke Nitz et al.
The Journal of clinical endocrinology and metabolism, 92(8), 3183-3188 (2007-06-15)
On the basis of its chromosomal localization and its role in the synthesis of the antilipolytic compound prostaglandin E(2), the prostaglandin E synthase 2 (PTGES2) is a candidate gene for type 2 diabetes. The aim of the present study was...
Inka Lindner et al.
Molecular nutrition & food research, 51(12), 1447-1451 (2007-11-06)
The prostaglandin E synthase 2 (PTGES2) gene maps to a locus linked to obesity and is involved in the synthesis of the antilipolytic compound prostaglandin E(2). In a recent study, we found an association of the minor PTGES2 Arg298His allele...

Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.

Entre em contato com a assistência técnica

Redes sociais

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

Pesquisa. Desenvolvimento. Produção.

Somos um fornecedor global líder para o setor de ciências biológicas, com soluções e serviços para pesquisa, desenvolvimento e produção de biotecnologia, além de produção e desenvolvimento de terapias farmacológicas com medicamentos.

© 2021 Merck KGaA, Darmstadt, Alemanha e/ou suas filiais. Todos os direitos reservados.

A reprodução não autorizada de quaisquer materiais deste site é estritamente proibida.