2050201
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
material
glass liner
descrição
Inlet Liner, Splitless, glass, for Varian
característica
straight
embalagem
pkg of 1 ea
manufacturer/tradename
Varian 01-900109-05 (OEM)
Varian 03-949437-90 (OEM, instrument manufacturer′s nondeactivated liner)
C × D.E. × D.I.
74 mm × 6.3 mm × 2.0 mm
compatibilidade
configured for
for use with Varian 1075/1077 Injectors
technique used for Trace analytes
Procurando produtos similares? Visit Guia de comparação de produtos
Descrição geral
The liner is the second most important part of the inlet. It is the centrepiece of the inlet system, basically a glass tube between the septum and the column, where the sample is evaporated and brought into the gas phase. Split/splitless inlet allows fast sample introduction and vaporization which is highly advantageous for small sample sizes. Though the injection modes for both split and splitless are carried out on the same inlet, there are certain differences like a splitless injection involves the complete introduction of the sample into the column whereas in a split injection, the sample is divided in the inlet as a result only part of the sample is introduced in the column.
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1075 and 1077 Injector)
Aplicação
Splitless inlet equipped with a flip-top sealing system in a thermal desorption unit maybe used to perform thermal desorption (TD) for the determination of pyrethroids in water samples.
Nota de rodapé
Instrument manufacturer′s nondeactivated liner.
Certificado de análise
Certificado de origem
Fire Debris Analysis.
Stauffer E
Science, 249-252 (2007)
Els Van Hoeck et al.
Journal of chromatography. A, 1157(1-2), 1-9 (2007-05-08)
Stir bar sorptive extraction (SBSE) was applied to the enrichment of pyrethroids in water samples. Analysis was done by retention time-locked GC/MS in selected ion-monitoring (SIM) mode using trans-permethrin-d(6) as internal standard. Thermal desorption (TD) was performed in a classical...