Merck
Analytical Vials
Glass vials with graduation.
Vials play a significant role in analytical analysis and result reproducibility. Vials must be inert and free of extractables or leachables to prevent affecting results. Using certified, application-specific, contaminant-free vials can significantly reduce risk. We offer a broad spectrum of Supelco® products including autosampler vials, general-purpose vials, and vial accessories for sample collection, storage, and processing. Supelco® vials are available in a variety of colors, sizes, volume capacities, and material compositions. Our vials and closures are designed, engineered, and manufactured for optimum performance in HPLC, GC/MS, LC/MS, and high-throughput analysis. In addition to specialized products, we offer:    
  • Caps, Crimp Seals & Stoppers
  • Headspace & SPME vials
  • Micro Reaction Vials
  • Glass and polyethylene scintillation vials
  • Polypropylene and polymethylpentene (TPX) vials
  • Polypropylene and glass microvolume inserts
  • Amber glass and PTFE storage containers
  • Silanized vials to improve polar compound recovery 
Products
2.0 (70)
4.0 (28)
0.3 (23)
1.5 (22)
40.0 (19)
20.0 (15)
glass (288)
polypropylene (133)
PTFE/silicone (115)
PTFE (54)
phenolic (47)
aluminum (42)
vials (277)
septa (106)
caps (40)
inserts (23)
bottles (11)
accessories (8)
Ordenar por relevância
Mostrando 1–20 de 494
Alterar visualização
Mostrando 1–20 de 494
Tampa de rosca magnética para frascos de headspace, rosca de 18 mm
SU860101
Tampa de rosca magnética para frascos de headspace, rosca de 18 mm
PTFE/silicone septum (white PTFE/tranparent blue silicone), septum thickness 1.3 mm, pkg of 100 ea
Frascos, tampa de rosca, vidro transparente (somente o frasco-ampola)
854190
Frascos, tampa de rosca, vidro transparente (somente o frasco-ampola)
volume 4 mL, clear glass vial, thread for 13-425, pkg of 100 ea
Headspace vial, screw top, rounded bottom (vial only)
SU860097
Headspace vial, screw top, rounded bottom (vial only)
volume 20 mL, clear glass vial, thread for 18, O.D. × H 22.5 mm × 75.5 mm, pkg of 100 ea
Certified Kits, screw thread vials, 12 x 32 mm, 9 mm thread, unassembled, pkg of 100
29378-U
Certified Kits, screw thread vials, 12 x 32 mm, 9 mm thread, unassembled, pkg of 100
volume 2 mL, clear glass vial, PTFE/silicone septum (bonded to cap)
Tampa de rosca, parte superior sólida com revestimento em PTFE, embalagem com 100
27141
Tampa de rosca, parte superior sólida com revestimento em PTFE, embalagem com 100
solid green melamine resin cap, F217/PTFE liner, for use with 4 mL vial with 13-425 thread
Frascos, tampa de rosca Thermoset verde-esmeralda com revestimento de PTFE, pré-montados, embalagem com 100 unidades
27181
Frascos, tampa de rosca Thermoset verde-esmeralda com revestimento de PTFE, pré-montados, embalagem com 100 unidades
volume 40 mL, clear glass vial, O.D. × H 29 mm × 82 mm, PTFE liner
Certified Vial Kit, Low Adsorption (LA), 2 mL, pk of 100
29651-U
Certified Vial Kit, Low Adsorption (LA), 2 mL, pk of 100
volume 2 mL, clear glass vial (with marking spot), natural PTFE/silicone septa, thread for 9 mm
Aluminum crimp seal (without septum), silver aluminum
27230-U
Aluminum crimp seal (without septum), silver aluminum
open center, diam. × opening 20 mm × 9.5 mm, pkg of 100 ea
Tampas de polipropileno certificadas com revestimento e com septos, pacote com 100
29320-U
Tampas de polipropileno certificadas com revestimento e com septos, pacote com 100
9 mm cap, PTFE/silicone septum (with slit), dark blue polypropylene cap
Tampas de polipropileno certificadas com revestimento e com septos, pacote com 100
29319-U
Tampas de polipropileno certificadas com revestimento e com septos, pacote com 100
9 mm cap, PTFE/silicone septum, dark blue polypropylene cap
Frascos, tampa de rosca, R.A.M.<SUP>™</SUP> (rosca de 9 mm), abertura grande, 12 x 32 mm
29007-U
Frascos, tampa de rosca, R.A.M. (rosca de 9 mm), abertura grande, 12 x 32 mm
volume 2 mL, clear glass vial (with graduated marking spot), pkg of × 100 ea
Aluminum crimp seal (without septum), silver aluminum
27016
Aluminum crimp seal (without septum), silver aluminum
tear-away, diam. 20 mm, pkg of 100 ea
Certified Kits, screw thread vials, 12 x 32 mm, 9 mm thread, unassembled, pkg of 100
29384-U
Certified Kits, screw thread vials, 12 x 32 mm, 9 mm thread, unassembled, pkg of 100
volume 2 mL, clear glass vial (with graduated marking spot), PTFE/silicone septum (with slit), (septa bonded to cap)
Frascos, tampa de rosca Thermoset verde-esmeralda com revestimento de PTFE, pré-montados, embalagem com 100 unidades
27138
Frascos, tampa de rosca Thermoset verde-esmeralda com revestimento de PTFE, pré-montados, embalagem com 100 unidades
volume 4 mL, clear glass vial, O.D. × H 15 mm × 45 mm, thread for 13-425
Tampa de rosca magnética para frascos de headspace, rosca de 18 mm
SU860103
Tampa de rosca magnética para frascos de headspace, rosca de 18 mm
PTFE/silicone septum (white PTFE/blue silicone), septum thickness 1.5 mm, pkg of 100 ea
Certified glass inserts for 12 x 32 mm, large opening vials
29435-U
Certified glass inserts for 12 x 32 mm, large opening vials
volume 0.2 mL, clear glass insert (with plastic bottom spring), O.D. × H 6 mm × 29 mm, pkg of 100 ea
Stoppers
27232
Stoppers
stopper diam. 20 mm, gray butyl rubber, pkg of 100 ea
Closures for Snap Ring vials
27427
Closures for Snap Ring vials
polypropylene seal, PTFE/silicone (with starburst), thread for 11 mm (snap ring), pkg of 100 ea
Frascos, tampa de rosca, vidro transparente (somente o frasco-ampola)
27151
Frascos, tampa de rosca, vidro transparente (somente o frasco-ampola)
volume 7 mL, clear glass vial, thread for 15-425, pkg of 100 ea
Tampa de rosca, parte superior sólida com revestimento em PTFE, embalagem com 100
27152
Tampa de rosca, parte superior sólida com revestimento em PTFE, embalagem com 100
green melamine resin solid cap, F217/PTFE liner, for use with 7 mL vial with 15-425 thread
Mass Spectrometry Vials
Mass Spectrometry Vials
Our certified, low adsorption (LA), LC/MS vials and kits are designed to reduce surface activity while improving analytical quantitation. The QSertVial™ microsampling device contains a fused, 300 μL glass insert and is compatible with most autosamplers. Our certified center drain (CD) vials and kits feature a unique conical interior bottom ideal for micro applications. Certified Supelco® vials, caps, and septa ensure minimal contamination and an air-tight seal.
Mininert® Valves
Mininert® Valves
Mininert® valves are highly dependable, leak-tight closures for screw top vials, crimp top vials, and other laboratory containers.
Precleaned Vials
Precleaned Vials
Our precleaned, Type 1 borosilicate clear or amber glass vials are washed in non-phosphate detergent, rinsed three times with tap water, and rinsed with ASTM Type-1 organic-free water to remove detergent. Post cleaning, vials are assembled with PTFE/silicone septa and open-top polypropylene caps.
VOA (EPA) Vials
VOA (EPA) Vials
VOA vials meet EPA standards for volatile organic sampling. The 40 mL vials are available in clear (Type 1, 33 expansion) and amber (Type 1, 51 expansion) borosilicate glass.