Merck
Filter Paper
Whatman® filter papers for qualitative and quantitative analyses
Used commonly in quantitative and qualitative analysis, filter papers must be closely matched to the application according to the physical characteristics of the filter paper. There are many sizes, materials, types, and grades of filter paper available for a variety of applications. Whatman® filter papers use only the highest quality raw materials for high reproducibility and high uniformity. Whatman® cellulose filter papers are manufactured from high-quality cotton liners that are treated to achieve an α-cellulose content >98%. A wide selection of Whatman® filter papers are available to suit numerous laboratory applications. Pre-pleated (folded) filter paper is also available for convenience.
Products
185.0 (51)
150.0 (49)
125.0 (46)
240.0 (43)
110.0 (35)
90.0 (30)
cellulose (432)
cotton (103)
paper (3)
cellulose fiber (2)
fibrous (1)
glass (1)
filter discs (563)
paper (4)
instruments (1)
Ordenar por relevância
Mostrando 1–20 de 577
Alterar visualização
Mostrando 1–20 de 577
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001090
Whatman® qualitative filter paper, Grade 1
circles, diam. 90 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001150
Whatman® qualitative filter paper, Grade 1
circles, diam. 150 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001125
Whatman® qualitative filter paper, Grade 1
circles, diam. 125 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001110
Whatman® qualitative filter paper, Grade 1
circles, diam. 110 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001070
Whatman® qualitative filter paper, Grade 1
circles, diam. 70 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper for technical use, Grade Shark Skin<SUP>®</SUP>, creped
WHA10347522
Whatman® qualitative filter paper for technical use, Grade Shark Skin®, creped
circles, diam. 340 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 4
WHA1004125
Whatman® qualitative filter paper, Grade 4
circles, diam. 125 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001055
Whatman® qualitative filter paper, Grade 1
circles, diam. 55 mm, pack of 100
Whatman<SUP>®</SUP> cellulose chromatography papers
WHA3030917
Whatman® cellulose chromatography papers
3MM Chr sheets, 46 x 57 cm, 100/pk
Whatman<SUP>®</SUP> quantitative filter paper, ashless, Grade 42
WHA1442125
Whatman® quantitative filter paper, ashless, Grade 42
circles, diam. 125 mm, pack of 100
Whatman<SUP>®</SUP> quantitative filter paper, ashless, Grade 41
WHA1441125
Whatman® quantitative filter paper, ashless, Grade 41
circles, diam. 125 mm, pack of 100
Whatman<SUP>®</SUP> quantitative filter paper, ashless, Grade 40
WHA1440150
Whatman® quantitative filter paper, ashless, Grade 40
circles, diam. 150 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 2
WHA1002125
Whatman® qualitative filter paper, Grade 2
circles, diam. 125 mm, pack of 100
Whatman<SUP>®</SUP> quantitative filter paper, ashless, Grade 40
WHA1440125
Whatman® quantitative filter paper, ashless, Grade 40
circles, diam. 125 mm, pack of 100
Whatman<SUP>®</SUP> quantitative filter paper, ashless, Grade 42
WHA1442150
Whatman® quantitative filter paper, ashless, Grade 42
circles, diam. 150 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 2
WHA1002150
Whatman® qualitative filter paper, Grade 2
circles, diam. 150 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001325
Whatman® qualitative filter paper, Grade 1
circles, diam. 25 mm, pack of 100
Whatman<SUP>®</SUP> quantitative filter paper, ashless, Grade 42
WHA1442042
Whatman® quantitative filter paper, ashless, Grade 42
circles, diam. 42.5 mm, pack of 100
Whatman<SUP>®</SUP> quantitative filter paper, ashless, Grade 42
WHA1442110
Whatman® quantitative filter paper, ashless, Grade 42
circles, diam. 110 mm, pack of 100
Whatman<SUP>®</SUP> qualitative filter paper, Grade 1
WHA1001047
Whatman® qualitative filter paper, Grade 1
circles, diam. 47 mm, pack of 100
Whatman® Qualitative Filter Paper
Qualitative filter papers are what most users would consider general purpose filter papers, designed for use in analytical separations and qualitative analytical techniques for the determination and identification of particles, contaminants, or material components. Qualitative papers are categorized by paper grades based on various properties including thickness, weight, and particle retention. The most common qualitative paper grades are 1-6, 595, 597, 598, and 602, which have applications across multiple industries.
Whatman® Quantitative Filter Paper
Quantitative filter papers are designed for sample preparation and gravimetric analysis, and are typically selected based upon the level of surface toughness and ash content required for the filtration procedure. Quantitative papers are acid treated to reduce ash, often to less than 0.01%. This low ash content is useful for quantitative analyses. After filtration or sample collection, the filter paper can be ignited and burned off, leaving very little residue that could interfere in downstream measurements.
  • Ashless filter paper: 0.007% ash nominal for grades 40 to 44 and a typical of 0.01% for 589 grades; very pure filters suitable for a wide range of critical analytical filtration procedures.
  • Hardened low ash filter paper: 0.015% ash nominal; treated with a strong acid to remove trace metals and produce high wet strength and chemical resistance; particularly suitable for Buchner filtration where the tough, smooth surface of the filter makes it easy to recover precipitates.
  • Hardened ashless filter paper: 0.005% ash nominal; acid hardened to give high wet strength and chemical resistance with extremely low ash content; tough surface makes these filters suitable for a wide range of critical filtration procedures.
Cytiva™ Blotting and Chromatography Paper
Whatman® 3MM Chr paper is the world’s most widely used blotting paper. A medium thickness paper (0.34 mm) is used extensively in electrophoresis for lifting of sequencing gels and relied upon by researchers doing Southern, Northern, and Western transfers. 3MM Chr paper is now available in the most widely used sizes. GB005 is a thick (1.5 mm) highly absorbent paper recommended for applications where fewer layers of blotting paper must still ensure a high capacity, including semi-dry blotting of proteins. GB003 is a general-purpose blotting paper (0.8 mm) made from pure raw materials with a high absorbency used as a membrane gel support. This thick paper is recommended for the lysis/denaturation of colony or plaque lifts and Western blots.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind. Ensures that no contamination will occur during transfer steps.
  • Manufactured and tested specifically for chromatographic and blotting techniques. This ensures the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting.
  • Whatman® 3MM Chr is considered the industry standard for blotting procedures.
  • Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes. Allows "out-of-the-box" usage and eliminates sheet-to-sheet variations.