Merck
FLAG® Purification
A close-up of a laboratory setting. The main focus is on a blue pipette dispensing a droplet into a small glass container partially filled with a pink-colored liquid. The background, although blurred, suggests the presence of various other laboratory instruments or containers.
FLAG® tags enable superior detection and robust purification of recombinant fusion proteins, with proven utility in numerous downstream applications from binding and activity assays to structural analysis. The FLAG® epitope tag is a short, hydrophilic, eight-amino acid peptide (DYKDDDDK tag) that is readily cleavable by enterokinase (EK). Due to its small size and hydrophilic nature, the FLAG® tag commonly resides on the surface of the fusion protein, minimizing effects on function, secretion, or transport of the fusion protein. Regardless of your final application, we have the tools needed to express, purify, and detect FLAG® fusion proteins. 
Products
Mostrando 1–20 de 26
Alterar visualização
Mostrando 1–20 de 26
Comparar
Número do produto
Nome do produto
Descrição do produto
Preços
Você visualizou 1–20 de 26 resultados
Página 1 de 2
A graphic representation featuring a pattern of interconnected purple circles and lines on a yellow background. The design resembles a simplified molecular structure or a network diagram, suggesting themes of interconnectivity or relationships.
FLAG® Tag Protein Expression
Our FLAG® tag protein expression portfolio includes vectors for efficient expression of recombinant fusion proteins in bacterial and mammalian systems. SnapFast™ vector technology allows you to easily move the gene-of-interest from one vector to another for more efficient and cost-effective cloning.
  • The standard FLAG® peptide (sequence: DYKDDDDK) is a small tag that can be incorporated with minimal risk of steric hindrance or negative impact on protein solubility
  • The 3xFLAG® tag sequence fuses three tandem FLAG® epitopes for enhanced (up to 200X) detection of fusion proteins
a vibrant yellow background with a centered purple square that has rounded corners. Inside the square, there is a symmetrical design with three horizontal arrows: one at the top pointing to the left and right, one in the middle pointing to both sides from the center, and one at the bottom mirroring the top arrow. A vertical line runs down from the top to bottom arrow through their points of symmetry. Scattered around these arrows are geometric shapes: circles and diamonds or rhombuses.
FLAG® Tag Protein Purification
We offer a variety of agarose affinity purification gels, magnetic beads, purification kits, and coated plates for isolation and purification of FLAG® tagged proteins.
  • Agarose affinity gels enable fusion-tagged protein purification using anti-FLAG® antibodies cross-linked to beaded agarose, and are suited for small to large scale purification, these resins are suitable for gravity flow column purification.
  • Anti-FLAG® M2 Magnetic Beads are used to purify recombinant fusion-tagged protein using a magnetic rack or platform to separate beads and isolate protein for rapid processing.
  • Anti-FLAG® High Sensitivity M2-coated 96-well plates are suitable for expression screening, the study of protein-protein interactions, and ELISA assays.
purple line art on a yellow background, resembling an abstract representation of the letter “K.” The design is simple with clean lines and no additional embellishments or textures.
FLAG® Tag Detection
FLAG® and 3xFLAG™ tags include binding sites for highly specific anti-FLAG® monoclonal antibodies as well as polyclonal antibodies and conjugates for ELISA, blotting applications, immunofluorescence, immunocytochemistry, immunohistochemistry (IHC), blotting, and flow cytometry.
  • The M2 anti-FLAG® antibody binds N-terminal FLAG®, C-terminal FLAG®, and Met-FLAG® fusion proteins.
  • The M1 anti-FLAG® antibody binding is calcium-dependent and specific for the N-terminus of the FLAG® tag.
  • The M5 anti-FLAG® antibody binds N-terminal FLAG® and Met-FLAG® fusion proteins.
 Monoclonal anti-FLAG® M1 antibody produced in mouseMonoclonal anti-FLAG® M2
antibody produced in Mouse
Monoclonal anti-FLAG® M5 antibody produced in Mouse
Catalog NumberF3040F1804F3165F4042
CloneM1M2M2M5
FormBuffered aqueous solutionBuffered aqueous glycerol solution (no sodium azide)Buffered aqueous solution (contains sodium azide)Buffered aqueous solution
Immunogen sequenceDYKDDDDKDYKDDDDKDYKDDDDKDYKDDDDK
Antibody purification methodPurified on Protein A SepharosePurified on proprietary FLAG® peptide-agarosePurified on Protein A SepharosePurified on Protein A Sepharose
KD53.4 nM9.3 nM9.3 nMUnavailable
SpecificityHighly specific. Recognizes only the FLAG® epitope when located at the free N-terminus.Recognizes the FLAG® sequence at N-terminus, Met-N-terminus,
C-terminus or at any internal site of the FLAG® fusion protein.
Strong recognition of the Met-N FLAG® fusion protein. Weak binding with N- or C-terminal fusion protein.
SensitivityDetects 1 ng of FLAG-BAP™ fusion protein on a dot blotDetects 2 ng of protein on a dot blotDetects <1 ng of Met-FLAG® fusion protein on a dot blot
Recommended for:Recommended for the detection of FLAG® fusion proteins in Mammalian, plants and bacterial expression systems. Very useful for affinity purification. Not recommended for detection of cytoplasmic expressed proteins.Western blot detection of target proteins expressed on E. coli,
plants or mammalian
crude cell lysate.
Western blot detection of target proteins expressed on mammalian and drosophilae cell lysate. Particularly useful for the detection of cytoplasmic expressed Met-FLAG fusion proteins. Not recommended for detection of fusion proteins expressed in E. coli
Applications*IP, IC, WB, EIAIP, IC, WB, EIAIP, IC, WB, EIAWB
Special requirementsBinding requires calcium. (Complex will dissociate in the absence of calcium ions)N/AN/AN/A
*IP=immunoprecipitation; IC=immunochemistry, WB=western blot, EIA=enzyme-linked immunosorbent assay (ELISA)
Antibody & binding characteristics
ANTI-FLAG M1
Mouse
IgG2b MAb
ANTI-FLAG M2
Mouse
IgG1 MAb
ANTI-FLAG
M5
Mouse
IgG1 MAb
Unprocessed N-terminal FLAG fusion protein:
Signal peptide - FLAG - Protein
-+Not determined
Met-N-terminal FLAG fusion protein:
Met - FLAG -Protein
-+++
N-terminal FLAG fusion protein:
FLAG - Protein
++Weak
Internal FLAG peptide:
Protein - FLAG - Protein
-+Not determined
C-terminal FLAG fusion protein:
Protein - FLAG
-+Weak
Calcium-dependent binding
(complex is dissociated in EDTA-containing buffer)
+--