Nome do produto
Indium beads, diam. 2-5 mm, 99.999% trace metals basis
Número do produto
264113
Marca do produto
aldrich
Número CAS
7440-74-6
Peso molecular
114.82
Teste
Especificação
Appearance (Color)Conforms to Requirements
Appearance (Form)Conforms
 Shot
Size2 - 5 mm
 Diameter
ICP Major AnalysisConfirmed
 Confirms Indium Component
Trace Metal Analysis≤20 ppm
PurityMeets Requirements
 99.999% Based on Trace Metals Analysis
Aluminum (Al)ppm
Arsenic (As)ppm
Boron (B)ppm
Barium (Ba)ppm
Calcium (Ca)ppm
Cobalt (Co)ppm
Chromium (Cr)ppm
Copper (Cu)ppm
Iron (Fe)ppm
Potassium (K)ppm
Lithium (Li)ppm
Magnesium (Mg)ppm
Manganese (Mn)ppm
Sodium (Na)ppm
Nickel (Ni)ppm
Lead (Pb)ppm
Antimony (Sb)ppm
Tin (Sn)ppm
Titanium (Ti)ppm
Vanadium (V)ppm
Zinc (Zn)ppm