Nome do produto
Sodium metabisulfite analytical standard
Número do produto
08982
Marca do produto
sial
Número CAS
7681-57-4
Peso molecular
190.11
Teste
Especificação
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
REDOX TITRATION≥ 98.0 %
REDOXTITRATION (METHOD)IODOMETRIC
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
CALCIUM (ICP)≤ 10 mg/kg
CADMIUM (ICP)≤ 5 mg/kg
COBALT (ICP)≤ 5 mg/kg
CHROMIUM (ICP)≤ 5 mg/kg
COPPER (ICP)≤ 5 mg/kg
IRON (ICP)≤ 5 mg/kg
POTASSIUM (ICP)≤ 200 mg/kg
MAGNESIUM (ICP)≤ 5 mg/kg
MANGANESE (ICP)≤ 5 mg/kg
NICKEL (ICP)≤ 5 mg/kg
LEAD (ICP)≤ 5 mg/kg
ZINC (ICP)≤ 5 mg/kg
ARSENIC TRACES (MHS-AAS)≤ 0.5 mg/kg
CHLORIDE (CL)≤ 50 mg/kg
THIOSULFATE (S2O3)≤ 500 mg/kg
EXPIRY PERIOD36 MONTHS