BR
PT
Aplicações
Produtos
Serviços
Suporte
Home
Search Results
Aquivion
®
D83-24B
(1)
Aquivion
®
D83-24B
Synonyms:
Aquivion
®
SO
3
H-form
Linear Formula:
(C
4
HF
7
O
4
S .C
2
F
4
)
x
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
802654
Aquivion
®
D83-24B, liquid, dispersion, 24% in water, PFSA eq. wt. 830 g/mole SO
3
H
Expand
View Pricing