BR
PT
Aplicações
Produtos
Serviços
Documentos
Suporte
Home
Search Results
Aquivion
®
E87-05S
(2)
Aquivion
®
E87-05S
Synonyms:
Aquivion
®
SO
3
H
Linear Formula:
[CF
2
CF(OCF
2
CF
2
SO
3
H)]
m
[CF
2
CF
2
]
n
CAS Number:
1163733-25-2
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-2 of 2
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
802719
Aquivion
®
E87-05S,
membrane sheet, contains CF
3
polymer chain ends as stabilizer, PFSA eq. wt. 870 g/mole SO
3
H, L × W × thickness 18 cm × 18 cm × 50 μm
Expand
View Pricing
802727
Aquivion
®
E87-05S,
membrane sheet, contains CF
3
polymer chain ends as stabilizer, PFSA eq. wt. 870 g/mole SO
3
H, L × W × thickness 31 cm × 31 cm × 50 μm
Expand
View Pricing
Page 1 of 1
Page 1 of 1