Merck
Atrazine solution 100 μg/mL in methanol, PESTANAL®, analytical standard

Atrazine solution

Atrazine solution 100 μg/mL in methanol, PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C8H14ClN5
CAS Number:
Molecular Weight:
215.68
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Atrazine solution, 1000 μg/mL in methyl tert-butyl ether, certified reference material
Atrazine solution, 100 μg/mL in methanol, PESTANAL®, analytical standard