Merck
D-Glucose (Dextrose) NIST® SRM® 917C

D-Glucose (Dextrose)

D-Glucose (Dextrose) NIST® SRM® 917C
Synonyms:
D-(+)-Glucose, Dextrose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
D-Glucose (Dextrose), NIST® SRM® 917C