Merck
NBS 28 (silicon and oxygen isotopes in silica sand) NIST® RM 8546

NBS 28 (silicon and oxygen isotopes in silica sand)

NBS 28 (silicon and oxygen isotopes in silica sand) NIST® RM 8546
Synonyms:
Sand, white quartz, Quartz, Silicon dioxide
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
NBS 28 (silicon and oxygen isotopes in silica sand), NIST® RM 8546