BR
PT
Aplicações
Produtos
Serviços
Suporte
Home
Search Results
Silica
(1)
Silica
Synonyms:
Silica, Silicon dioxide, Silicic anhydride, Silicon dioxide amorphous, Silica, fumed
Linear Formula:
SiO
2
CAS Number:
112945-52-5
Molecular Weight:
60.08
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
381276
Silica, 99.8%
Expand
View Pricing