BR
PT
Aplicações
Produtos
Serviços
Suporte
Home
Search Results
Enantioselective synthesis of (R)-phenylephrine hydrochloride
Enantioselective synthesis of (R)-phenylephrine hydrochloride
Tetrahedron Letters (2003)
Pandey RK, et al.