  • Search Results
  • Dysregulation of mitochondrial quality control processes contribute to sarcopenia in a mouse model of premature aging
Dysregulation of mitochondrial quality control processes contribute to sarcopenia in a mouse model of premature aging
Testing (2013)
Joseph A M,et al.