Effects of zinc oxide nanoparticles and zinc sulfate on the testis of common carp, Cyprinus carpio.
Nanotoxicology (2019-01-22)
Seetharam Deepa, Raju Murugananthkumar, Yugantak Raj Gupta, Manjunatha Gowda K S, Balasubramanian Senthilkumaran
RESUMO
The present study analyzed the effects of zinc oxide nanoparticles (ZnO-NPs) and zinc sulfate (ZnSO4) in the testis of six-month-old common carp Cyprinus carpio exposed to three different doses, viz., 10, 50, and 100 μg/L for 21 days. Characterization of ZnO-NPs was done after sonication, the size and shape of ZnO-NPs were determined as ∼20-30 nm spherical structure measured zeta potential of +26.0 mV. After treatment, determination of zinc (Zn) concentration in the testes revealed desired impact of the exposure. Expression of several transcription factors and few steroidogenic enzyme genes in the treated testis showed significant downregulation than the control. Measurement of oxidative stress-related enzymes such as catalase, superoxide dismutase, and glutathione-S-transferase revealed substantial elevation in the testis of treated groups when compared to control. Histological analysis of testis exhibited dose-related response, defective lumen, and slow progression of spermatogenesis. Exposure of both the forms of Zn on TM3 Leydig cell culture displayed loss of adhesion, clumping with decreased viability, and a significant increase in the apoptotic cells. In addition, comet and intracellular reactive oxygen species (ROS) assays authenticated DNA damage upon treatment with a significant increase in ROS. Histological analysis after treatment withdrawal showed revival of testis in carp to rescue the effect. Thus, the present report highlights the adverse effect of Zn on the testis function in common carp as well as evident drastically toxic in in vitro cultures.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, pellets, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Paraplast Plus®, for tissue embedding
Sigma-Aldrich
3,4-Ethylenedioxypyrrole, 2 % (w/v) in THF