Merck
  • Início
  • Resultados da busca
  • Vector competence analysis of two Aedes aegypti lineages from Bello, Colombia, reveals that they are affected similarly by dengue-2 virus infection.
Vector competence analysis of two Aedes aegypti lineages from Bello, Colombia, reveals that they are affected similarly by dengue-2 virus infection.
Archives of virology (2018-10-10)
Laura Silvana Pérez-Restrepo, Omar Triana-Chávez, Ana María Mejía-Jaramillo, Sair Orieta Arboleda-Sánchez
RESUMO
Dengue is the second most prevalent vector-borne disease after malaria in Colombia. It is caused by dengue virus, an arbovirus that exhibits high epidemic power, which is evidenced by its occurrence in more than 80% of the country, largely because of the extensive dispersion of the mosquito vector Aedes aegypti. The existence of two lineages of Ae. aegypti has been proposed based on genetic differences at the mitochondrial level, and they have been reported to circulate in similar proportions in the municipality of Bello (Colombia). It has been suggested that the differentiation of these lineages could influence features such as vector competence (VC) and life table. With the aim of testing this hypothesis, female mosquitoes from both lineages collected from Bello were orally challenged with dengue virus serotype 2 (strain D2-HAN) to measure infection, dissemination, survival and fecundity. Analysis of VC showed an increase in viral titer over time; however, no significant differences were observed between the lineages. The survival rate was not different between the infected lineages, but comparing lineages, it was lower in infected mosquitoes, which may affect the intensity of transmission. Finally, we conclude that the genetic differentiation of Ae. aegypti into lineages did not confer differences in epidemiological status when the mosquitoes were infected with this D2 serotype strain.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture