• Início
  • Resultados da busca
  • Epigenetic regulation of microglial phosphatidylinositol 3-kinase pathway involved in long-term potentiation and synaptic plasticity in rats.
Epigenetic regulation of microglial phosphatidylinositol 3-kinase pathway involved in long-term potentiation and synaptic plasticity in rats.
Glia (2019-11-09)
Genevieve Saw, Kumar Krishna, Neelima Gupta, Tuck Wah Soong, Karthik Mallilankaraman, Sreedharan Sajikumar, S Thameem Dheen
RESUMO
Microglia are the main form of immune defense in the central nervous system. Microglia express phosphatidylinositol 3-kinase (PI3K), which has been shown to play a significant role in synaptic plasticity in neurons and inflammation via microglia. This study shows that microglial PI3K is regulated epigenetically through histone modifications and posttranslationally through sumoylation and is involved in long-term potentiation (LTP) by modulating the expression of brain-derived neurotrophic factor (BDNF), which has been shown to be involved in neuronal synaptic plasticity. Sodium butyrate, a histone deacetylase inhibitor, upregulates PI3K expression, the phosphorylation of its downstream effectors, AKT and cAMP response element-binding protein (CREB), and the expression of BDNF in microglia, suggesting that BDNF secretion is regulated in microglia via epigenetic regulation of PI3K. Further, knockdown of SUMO1 in BV2 microglia results in a decrease in the expression of PI3K, the phosphorylation of AKT and CREB, as well as the expression of BDNF. These results suggest that microglial PI3K is epigenetically regulated by histone modifications and posttranslationally modified by sumoylation, leading to altered expression of BDNF. Whole-cell voltage-clamp showed the involvement of microglia in neuronal LTP, as selective ablation or disruption of microglia with clodronate in rat hippocampal slices abolished LTP. However, LTP was rescued when the same hippocampal slices were treated with active PI3K or BDNF, indicating that microglial PI3K/AKT signaling contributes to LTP and synaptic plasticity. Understanding the mechanisms by which microglial PI3K influences synapses provides insights into the ways it can modulate synaptic transmission and plasticity in learning and memory.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin solution from bovine pancreas, BioReagent, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium butyrate, 98%