Merck
  • Início
  • Resultados da busca
  • EGFR inhibitor AG1478 blocks the formation of 3D structures mainly through ERK signaling pathway in Matrigel-induced 3D reconstruction of eccrine sweat gland-like structures.
EGFR inhibitor AG1478 blocks the formation of 3D structures mainly through ERK signaling pathway in Matrigel-induced 3D reconstruction of eccrine sweat gland-like structures.
Journal of molecular histology (2020-03-29)
Liyun Chen, Lijie Du, Lei Zhang, Sitian Xie, Xiang Zhang, Haihong Li
RESUMO
EGFR signaling plays important roles in the development of eccrine sweat glands. We previously demonstrate that Matrigel induces eccrine sweat gland cells to reconstruct the three-dimensional (3D) structures of eccrine sweat glands, but the mechanisms are still unknown. In the study, eccrine sweat gland cells were cultured within a 3D Matrigel, and EGFR inhibitor AG1478, or MEK1/2 inhibitor U0126, were added to the medium respectively. The morphology of the 3D-reconstructed eccrine sweat gland-like structures was observed, the localization of phospho-EGFR was detected, and protein levels of EGFR, phospho-EGFR, phospho-JAK, phospho-AKT and phospho-ERK were examined. The results showed that cells treatment with AG1478 from Day 0 of 3D cultures blocked formation of spheroid-like structures. AG1478 administration caused reduced phospho-EGFR, concomitant with downregulation of phospho-ERK1/2, but not phospho-JAK or phospho-AKT. Phospho-EGFR and phospho-ERK were reduced, and only a small number of 3D-structures were formed following treatment with U0126. We conclude that EGFR plays important roles in Matrigel-induced 3D structures of eccrine sweat gland-like structures, and ERK1/2 signaling is responsible, at least in part, for the effect of EGFR.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tyrphostin AG 1478, ≥98%
Sigma-Aldrich
2-Phenylindole, technical grade, 95%