  • Search Results
  • The essential role of fructose-1,6-bisphosphatase 2 enzyme in thermal homeostasis upon cold stress.
The essential role of fructose-1,6-bisphosphatase 2 enzyme in thermal homeostasis upon cold stress.
Experimental & molecular medicine (2020-03-24)
Hyun-Jun Park, Hye Rim Jang, Shi-Young Park, Young-Bum Kim, Hui-Young Lee, Cheol Soo Choi
ABSTRACT
Skeletal muscle is a major organ for glucose disposal and thermogenesis. While hepatic fructose-1,6-bisphosphatase is well known as a key enzyme for gluconeogenesis, the role of muscle fructose-1,6-bisphosphatase 2 (Fbp2) in glucose disposal and thermogenesis is unknown. Here, using Fbp2 knockout (KO) mice, we assessed the physiological role of Fbp2 in energy and glucose metabolism and thermogenesis. In vivo assessments of energy metabolism, glucose metabolism, and thermogenesis were performed by indirect calorimetry, hyperinsulinemic-euglycemic clamp, and cold challenge studies, respectively. Under both feeding and fasting conditions, Fbp2 KO mice showed similar phenotypes regarding energy and glucose metabolism compared to wild-type (WT) mice. However, Fbp2 KO mice were severely intolerant to cold challenge under fasting conditions. Mechanistically, the cold-induced intramuscular conversion of lactate to glycogen (glyconeogenesis) is completely abolished in the KO muscle, which leads to a lack of glycogen source for thermogenesis in Fbp2 KO mice. The cold-intolerant phenotype of KO mice disappeared after feeding, and the KO mice were equally as cold tolerant as the WT mice and survived during the cold challenge for three weeks. Taken together, these data demonstrate that Fbp2 is essential for muscle thermogenesis by replenishing the intramuscular glycogen pool through glyconeogenesis when the exogenous glucose source is limited. These data imply the physiological importance of Fbp2 in thermal homeostasis and suggest a potential novel therapy targeted to increase glycogen replenishment upon cold stress.
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Isoprenaline hydrochloride
Sigma-Aldrich
Glucose (GO) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Sodium L-lactate, ~98%
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules