• Início
  • Resultados da busca
  • Thrombin generation test with the calibrated automated thrombogram and anticoagulant activity of Mentha crispa.
Thrombin generation test with the calibrated automated thrombogram and anticoagulant activity of Mentha crispa.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis (2019-10-24)
Paula M Leite, Ana P N Miranda, Juliana M Amorim, Rita C F Duarte, André A G Faraco, Maria D G Carvalho, Rachel O Castilho
RESUMO
: Evaluate the in-vitro effect of Mentha crispa extract on blood coagulation, compare the conventional coagulometric tests with thrombin generation test (TGT), and study the qualitative micromolecular composition of M. crispa. Extract of M. crispa was incubated with plasma and used in the coagulometric tests: prothrombin and activated partial thromboplastin times, fibrinogen, and TGT. A phytochemical prospection was performed to evaluate the chemical composition of this extract. The extract was efficient in prolonging prothrombin time and activated partial thromboplastin time, and reducing fibrinogen levels and TGT parameters, indicating that the extract of M. crispa inhibited the intrinsic and extrinsic pathways of blood coagulation. The results obtained in TGT are in agreement with the results of conventional coagulometric tests and the in-vitro anticoagulant activity of M. crispa suggests that its use by patients using oral anticoagulants deserves caution.
MATERIAIS
Número do produto
Marca
Descrição do produto
Rutin, phyproof® Reference Substance