Flavonoids and antioxidant activity of rare and endangered fern: Isoetes sinensis.
PloS one (2020-05-13)
Xin Wang, Guohua Ding, Baodong Liu, Quanxi Wang
RESUMO
Isoetes sinensis Palmer is a critically endangered, first-class protected plant in China. Until now, researchers have primarily focused on the ultrastructure, phylogeny, and transcriptomes of the plant. However, flavonoid profiles and bioactivity of I. sinensis have not been extensively investigated. To develop the endangered I. sinensis for edible and medicinal purposes, flavonoid content, chemical constitution, and antioxidant activities were investigated in this study. Results revealed the following. 1) The total flavonoid content was determined as 10.74 ± 0.25 mg/g., 2) Antioxidant activities were stronger than most ferns, especially ABTS free radical scavenging activities. 3) Four flavones, containing apigenin, apigenin-7-glucuronide, acacetin-7-O-glcopyranoside, and homoplantageninisoetin; four flavonols, namely, isoetin, kaempferol-3-O-glucoside, quercetin-3-O-[6"-O-(3-hydroxy-3-methylglutaryl)-β-D-glucopyranoside], and limocitrin-Neo; one prodelphinidin (procyanidins;) and one nothofagin (dihydrochalcone) were tentatively identified in the mass spectrometry-DAD (254nm) chromatograms. This study was the first to report on flavonoid content and antioxidant activities of I. sinensis. Stronger antioxidant activity and flavonoid content suggests that the endangered I. sinensis is an important and potentially edible and medicinal plant.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Phenazine methosulfate, ≥90% (UV)
Rutin, phyproof® Reference Substance